Learn More
Medchemexpress LLC HY-16500 5mg Medchemexpress, Tolrestat CAS:82964-04-3 Purity:>98%
SDP
Catalog No. 502081888
Encompass_Preferred
Medchemexpress, HY-16500 5mg Tolrestat CAS:82964-04-3 Tolrestat is a potent, orally active aldose reductase inhibitor with IC50 of 35 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1888 Supplier Medchemexpress LLC Supplier No. HY165005MG
Medchemexpress, HY-16500 5mg Tolrestat CAS:82964-04-3 Tolrestat is a potent, orally active aldose reductase inhibitor with IC50 of 35 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.