Learn More
Apexbio Technology LLC Tolterodine tartrate 124937-52-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092645
Encompass_Preferred
Tolterodine tartrate (CAS 124937-52-6) is the tartrate salt form of tolterodine a small molecule that acts as a competitive antagonist at muscarinic acetylcholine receptors By selectively inhibiting these receptors particularly subtypes involved in urinary bladder contraction tolterodine tartrate reduces detrusor muscle activity This mechanism underlies its utility in studies related to bladder function muscarinic receptor pharmacology and disorders characterized by overactive bladder The compound is commonly employed in preclinical and clinical research investigating the modulation of cholinergic signaling in urological and neurological contexts
Catalog No. 50-000-92645 Supplier Apexbio Technology LLC Supplier No. B16205.1
Tolterodine tartrate (CAS 124937-52-6) is the tartrate salt form of tolterodine a small molecule that acts as a competitive antagonist at muscarinic acetylcholine receptors By selectively inhibiting these receptors particularly subtypes involved in urinary bladder contraction tolterodine tartrate reduces detrusor muscle activity This mechanism underlies its utility in studies related to bladder function muscarinic receptor pharmacology and disorders characterized by overactive bladder The compound is commonly employed in preclinical and clinical research investigating the modulation of cholinergic signaling in urological and neurological contexts