Learn More
Medchemexpress LLC Tolterodine tartrate | 124937-52-6 | 98.6% | C26H37NO7 | 1 ML
SDP
Catalog No. 5000433493
Encompass_Preferred
Tolterodine tartrate is a mAChR inhibitor and a substrate for cytochrome P450 enzymes. It competitively binds acetylcholine, which reduces sympathetic excitation and inhibits involuntary bladder muscle contraction. It restores the Nrf2/NF-κB signaling pathway, providing protection against inflammatory response and ferroptosis, and ameliorates LPS-induced reactive oxygen species production and lipid oxidation. This product can be used for research related to urinary tract infections and overactive bladder.
  • Competitively binds acetylcholine
  • Reduces sympathetic excitation
  • Inhibits involuntary bladder muscle contraction
  • Restores Nrf2/NF-κB signaling pathway
  • Provides protection against inflammatory response and ferroptosis
  • Ameliorates LPS-induced reactive oxygen species production and lipid oxidation
Catalog No. 50-004-33493 Supplier Medchemexpress LLC Supplier No. HY9001010MM/1ML
Tolterodine tartrate is a mAChR inhibitor and a substrate for cytochrome P450 enzymes. It competitively binds acetylcholine, which reduces sympathetic excitation and inhibits involuntary bladder muscle contraction. It restores the Nrf2/NF-κB signaling pathway, providing protection against inflammatory response and ferroptosis, and ameliorates LPS-induced reactive oxygen species production and lipid oxidation. This product can be used for research related to urinary tract infections and overactive bladder.
  • Competitively binds acetylcholine
  • Reduces sympathetic excitation
  • Inhibits involuntary bladder muscle contraction
  • Restores Nrf2/NF-κB signaling pathway
  • Provides protection against inflammatory response and ferroptosis
  • Ameliorates LPS-induced reactive oxygen species production and lipid oxidation