Learn More
Medchemexpress LLC Tolterodine (Tartrate) | 124937-52-6 | 98.6% | C26H37NO7 | 25 MG
SDP
Catalog No. 5000357850
Encompass_Preferred
Tolterodine tartrate is an mAChR inhibitor and a substrate for cytochrome P450 enzymes. It competitively binds acetylcholine, reducing sympathetic excitation and inhibiting involuntary bladder muscle contraction. It also restores the Nrf2/NF-κB signaling pathway, protecting against inflammatory response and ferroptosis. It is used for research on urinary tract infections and overactive bladder.
  • mAChR inhibitor
  • Substrate for cytochrome P450 enzymes
  • Competitively binds acetylcholine
  • Reduces sympathetic excitation
  • Inhibits involuntary bladder muscle contraction
  • Restores Nrf2/NF-κB signaling pathway
  • Protects against inflammatory response and ferroptosis
  • Ameliorates LPS-induced oxidative stress
  • Research use for urinary tract infections and overactive bladder
Catalog No. 50-003-57850 Supplier Medchemexpress LLC Supplier No. 25MGHY90010
Tolterodine tartrate is an mAChR inhibitor and a substrate for cytochrome P450 enzymes. It competitively binds acetylcholine, reducing sympathetic excitation and inhibiting involuntary bladder muscle contraction. It also restores the Nrf2/NF-κB signaling pathway, protecting against inflammatory response and ferroptosis. It is used for research on urinary tract infections and overactive bladder.
  • mAChR inhibitor
  • Substrate for cytochrome P450 enzymes
  • Competitively binds acetylcholine
  • Reduces sympathetic excitation
  • Inhibits involuntary bladder muscle contraction
  • Restores Nrf2/NF-κB signaling pathway
  • Protects against inflammatory response and ferroptosis
  • Ameliorates LPS-induced oxidative stress
  • Research use for urinary tract infections and overactive bladder