Learn More
Medchemexpress LLC Tolterodine (tartrate) | 124937-52-6 | MFCD00045558 | 98.6% | C26H37NO7 | 250 MG
SDP
Catalog No. 5000358421
Encompass_Preferred
Tolterodine tartrate is an mAChR inhibitor and substrate for cytochrome P450 enzymes, used in research for urinary tract infections and overactive bladder. It competitively binds acetylcholine, reducing sympathetic excitation and inhibiting involuntary bladder muscle contraction.
  • Inhibits muscarinic acetylcholine receptors
  • Substrate for cytochrome P450 enzymes
  • Reduces sympathetic excitation
  • Inhibits involuntary bladder muscle contraction
  • Restores Nrf2/NF-κB signaling pathway
  • Protects against inflammatory response and ferroptosis
  • Ameliorates LPS-induced oxidative stress
Catalog No. 50-003-58421 Supplier Medchemexpress LLC Supplier No. 250MGHY90010
Tolterodine tartrate is an mAChR inhibitor and substrate for cytochrome P450 enzymes, used in research for urinary tract infections and overactive bladder. It competitively binds acetylcholine, reducing sympathetic excitation and inhibiting involuntary bladder muscle contraction.
  • Inhibits muscarinic acetylcholine receptors
  • Substrate for cytochrome P450 enzymes
  • Reduces sympathetic excitation
  • Inhibits involuntary bladder muscle contraction
  • Restores Nrf2/NF-κB signaling pathway
  • Protects against inflammatory response and ferroptosis
  • Ameliorates LPS-induced oxidative stress