Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolterodine Tartrate Pharmaceutical Secondary Standard: Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111018333 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8333 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8333 Supplier MilliporeSigma Supelco Supplier No. PHR1891500MG
Add to Cart
Add to Cart

Tolterodine Tartrate is a tertiary muscarinic antagonist, basically used to manage urinary frequency, urgency, and incontinence in detrusor instability.

TRUSTED_SUSTAINABILITY

Specifications

CAS 124937-52-6
Linear Formula C22H31 NO · C4H6O6
Synonym (R)-Tolterodine L-tartrate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 475.57
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.