Learn More
Apexbio Technology LLC Toltrazuril 69004-03-1 1g
SDP
Catalog No. 5000092889
Encompass_Preferred
Toltrazuril (69004-03-1) is an antiprotozoal compound targeting coccidial parasites It is designed to interfere with mitochondrial energy metabolism and the division of protozoan parasites thereby inhibiting parasite proliferation Toltrazuril exerts its biological activity primarily through disruption of mitochondrial function in protozoa In in vitro studies toltrazuril demonstrates inhibitory activity against coccidial parasites with reported IC50 values typically ranging from 0 02 to 0 3 g/mL depending on testing conditions and parasite species Based on these pharmacological properties toltrazuril holds research potential in experimental investigations of antiprotozoal mechanisms parasite-host interactions and therapeutic evaluations against protozoan pathogens
Catalog No. 50-000-92889 Supplier Apexbio Technology LLC Supplier No. B20541000
Toltrazuril (69004-03-1) is an antiprotozoal compound targeting coccidial parasites It is designed to interfere with mitochondrial energy metabolism and the division of protozoan parasites thereby inhibiting parasite proliferation Toltrazuril exerts its biological activity primarily through disruption of mitochondrial function in protozoa In in vitro studies toltrazuril demonstrates inhibitory activity against coccidial parasites with reported IC50 values typically ranging from 0 02 to 0 3 g/mL depending on testing conditions and parasite species Based on these pharmacological properties toltrazuril holds research potential in experimental investigations of antiprotozoal mechanisms parasite-host interactions and therapeutic evaluations against protozoan pathogens