Learn More
Cayman Chemical ToltrazurIl sulfon 10mg
SDP
Catalog No. 502991819
Encompass_Preferred
An active metabolite of toltrazuril; formed from toltrazuril via the intermediate metabolite toltrazuril sulfoxide by CYP enzymes, including CYP3A; inhibits T. gondii tachyzoite production in African green monkey kidney cells at 0.1, 1, and 5 UG/ml; completely prevents infection with T. gondii tachyzoites in a mouse model of fatal toxoplasmosis at 10 or 20 mg/kg one day prior to, and daily for 10 days following, infection; reduces the number of infected mice in the same model at 10 mg/kg for 11 days starting six days following infection
Catalog No. 50-299-1819 Supplier Cayman Chemical Supplier No. 2965510 MG
An active metabolite of toltrazuril; formed from toltrazuril via the intermediate metabolite toltrazuril sulfoxide by CYP enzymes, including CYP3A; inhibits T. gondii tachyzoite production in African green monkey kidney cells at 0.1, 1, and 5 UG/ml; completely prevents infection with T. gondii tachyzoites in a mouse model of fatal toxoplasmosis at 10 or 20 mg/kg one day prior to, and daily for 10 days following, infection; reduces the number of infected mice in the same model at 10 mg/kg for 11 days starting six days following infection