Learn More
TARGETMOL CHEMICALS INC Toltrazuril sulfone 100MG
SDP
Catalog No. 502876675
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Toltrazuril sulfone (Ponazuril) is an antiprotozoal agent that acts upon Coccidia parasites. Purity 98.37%
Catalog No. 50-287-6675 Supplier TARGETMOL CHEMICALS INC Supplier No. T5052100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Toltrazuril sulfone (Ponazuril) is an antiprotozoal agent that acts upon Coccidia parasites. Purity 98.37%