Learn More
Medchemexpress LLC Toltrazuril (sulfone) | 69004-04-2 | 98.6% | 457.38 | 1 G
SDP
Catalog No. 5000391282
Encompass_Preferred
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril that exhibits antiprotozoal activity. It is a triazine anticoccidial developed for the prevention of coccidiosis in poultry. The product is for research use only and not for sale to patients.
  • Antiprotozoal activity.
  • Metabolite of Toltrazuril.
  • Triazine anticoccidial.
  • Developed to prevent coccidiosis in poultry.
  • Inhibits the development of merozoites of *S. neurona* *in vitro*.
  • Inhibits the development of *N. caninum* *in vitro* after approximately 48 hours post-exposure.
  • Exhibits inhibitory effects possibly by targeting different enzyme/enzyme systems in various apicomplexans.
  • Inhibits *T. gondii* replication *in vitro* after the second division by endodyogeny.
  • Effective *in vivo* in preventing and treating toxoplasmosis in mice at dosages of 10-20 mg/kg (p.o.; daily; for 10 days).
Catalog No. 50-003-91282 Supplier Medchemexpress LLC Supplier No. HY170081G
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril that exhibits antiprotozoal activity. It is a triazine anticoccidial developed for the prevention of coccidiosis in poultry. The product is for research use only and not for sale to patients.
  • Antiprotozoal activity.
  • Metabolite of Toltrazuril.
  • Triazine anticoccidial.
  • Developed to prevent coccidiosis in poultry.
  • Inhibits the development of merozoites of *S. neurona* *in vitro*.
  • Inhibits the development of *N. caninum* *in vitro* after approximately 48 hours post-exposure.
  • Exhibits inhibitory effects possibly by targeting different enzyme/enzyme systems in various apicomplexans.
  • Inhibits *T. gondii* replication *in vitro* after the second division by endodyogeny.
  • Effective *in vivo* in preventing and treating toxoplasmosis in mice at dosages of 10-20 mg/kg (p.o.; daily; for 10 days).