Learn More
Medchemexpress LLC Toltrazuril (sulfone) | 69004-04-2 | 98.6% | 457.38 | 5 G
SDP
Catalog No. 5000391232
Encompass_Preferred
Toltrazuril sulfone (Ponazuril) is a metabolite that exhibits antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry.
  • Inhibits the development of merozoites of *S. neurona*.
  • Inhibits the development of *N. caninum* after approximately 48 hours post-exposure.
  • Exhibits inhibitory effects possibly by targeting different enzyme/enzyme systems in different apicomplexans.
  • Inhibits *T. gondii* replication after the second division by endodyogeny.
  • Effective in preventing and treating toxoplasmosis in mice.
Catalog No. 50-003-91232 Supplier Medchemexpress LLC Supplier No. HY170085G
Toltrazuril sulfone (Ponazuril) is a metabolite that exhibits antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry.
  • Inhibits the development of merozoites of *S. neurona*.
  • Inhibits the development of *N. caninum* after approximately 48 hours post-exposure.
  • Exhibits inhibitory effects possibly by targeting different enzyme/enzyme systems in different apicomplexans.
  • Inhibits *T. gondii* replication after the second division by endodyogeny.
  • Effective in preventing and treating toxoplasmosis in mice.