Learn More
Medchemexpress LLC Toltrazuril sulfone | 69004-04-2 | 98.6% | 457.38 | 5 MG
SDP
Catalog No. 5000390706
Encompass_Preferred
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril with antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry.
  • Antiprotozoal activity.
  • Inhibits the development of merozoites of S. neurona.
  • Inhibits the development of N. caninum after approximately 48 hours post-exposure.
  • Exhibits inhibitory action by possibly targeting different enzyme/enzyme systems in various apicomplexans.
  • Inhibits T. gondii replication after the second division by endodyogeny.
  • Effective in preventing and treating toxoplasmosis in mice.
Catalog No. 50-003-90706 Supplier Medchemexpress LLC Supplier No. HY170085MG
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril with antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry.
  • Antiprotozoal activity.
  • Inhibits the development of merozoites of S. neurona.
  • Inhibits the development of N. caninum after approximately 48 hours post-exposure.
  • Exhibits inhibitory action by possibly targeting different enzyme/enzyme systems in various apicomplexans.
  • Inhibits T. gondii replication after the second division by endodyogeny.
  • Effective in preventing and treating toxoplasmosis in mice.