Learn More
Medchemexpress LLC Toltrazuril sulfone | 69004-04-2 | 98.6% | 457.38 | 50 MG
SDP
Catalog No. 5000391256
Encompass_Preferred
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril with antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry. It is intended for research use only.
  • Antiprotozoal activity
  • Anticoccidial (triazine)
  • Developed to prevent coccidiosis in poultry
  • Inhibits the development of merozoites of *S. neurona* in vitro
  • Inhibits the development of *N. caninum* after approximately 48 hours post-exposure in vitro
  • Exhibits inhibitory effects, possibly by targeting different enzyme/enzyme systems in various apicomplexans in vitro
  • Inhibits *T. gondii* replication after the second division by endodyogeny in vitro
  • Effective in preventing and treating toxoplasmosis in mice in vivo
Catalog No. 50-003-91256 Supplier Medchemexpress LLC Supplier No. HY1700850MG
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril with antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry. It is intended for research use only.
  • Antiprotozoal activity
  • Anticoccidial (triazine)
  • Developed to prevent coccidiosis in poultry
  • Inhibits the development of merozoites of *S. neurona* in vitro
  • Inhibits the development of *N. caninum* after approximately 48 hours post-exposure in vitro
  • Exhibits inhibitory effects, possibly by targeting different enzyme/enzyme systems in various apicomplexans in vitro
  • Inhibits *T. gondii* replication after the second division by endodyogeny in vitro
  • Effective in preventing and treating toxoplasmosis in mice in vivo