Learn More
Medchemexpress LLC Toltrazuril (sulfone) | 69004-04-2 | 98.6% | 457.38 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY17008500MG
Encompass_Preferred
Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril, possessing antiprotozoal activity. It is a triazine anticoccidial developed to prevent coccidiosis in poultry.
  • Inhibits the development of merozoites of *S. neurona*.
  • It inhibits the development of *N. caninum* approximately 48 hours post-exposure.
  • It exhibits inhibitory effects possibly by targeting different enzyme/enzyme systems in various apicomplexans.
  • Inhibits *T. gondii* replication after the second division by endodyogeny (5 mg/ml; 20 hours).
  • Effective in preventing and treating toxoplasmosis in mice (10-20 mg/kg; p.o.; daily; for 10 days).
Catalog No. 50-003-91245