Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Toltrazuril sulfoxide | 69004-15-5 | MFCD10567359 | 99.6% | 441.38 | C18H14F3N3O5S | 5 MG
SDP

Catalog No. 5000222519
Encompass_Preferred

Toltrazuril sulfoxide is the short-lived intermediary metabolite of toltrazuril and is supplied as an analytical standard for research and analytical applications. It is intended for in vitro use and method development, offering defined storage and solubility properties for consistent handling and reproducible results.

  • Analytical standard for research and method development.
  • Short-lived metabolite relevant to metabolic and pharmacokinetic studies.
  • High purity (99.6%) for accurate analytical results.
  • Soluble in DMSO at 250 mg/mL; ultrasonic aid recommended.
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-22519 Supplier Medchemexpress LLC Supplier No. HY1364385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Toltrazuril sulfoxide is the short-lived intermediary metabolite of toltrazuril and is supplied as an analytical standard for research and analytical applications. It is intended for in vitro use and method development, offering defined storage and solubility properties for consistent handling and reproducible results.

  • Analytical standard for research and method development.
  • Short-lived metabolite relevant to metabolic and pharmacokinetic studies.
  • High purity (99.6%) for accurate analytical results.
  • Soluble in DMSO at 250 mg/mL; ultrasonic aid recommended.
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.