Learn More
Medchemexpress LLC Toltrazuril sulfoxide | 69004-15-5 | MFCD10567359 | 99.6% | C18H14F3N3O5S | 10MG
SDP
Catalog No. 5000260951
Encompass_Preferred
Toltrazuril sulfoxide is an analytical standard corresponding to the short-lived sulfoxide metabolite of toltrazuril. It is intended for research and analytical applications, including metabolic studies, method development, and quantitative analysis of toltrazuril metabolites.
  • Analytical standard suitable for metabolic and pharmacokinetic studies.
  • High purity (99.6%) for reliable analytical results.
  • Provided in small pack sizes for trace-level assays and method validation.
  • Compatible with LC-MS and other chromatographic detection methods.
  • Stable solid form for routine storage and handling.
Catalog No. 50-002-60951 Supplier Medchemexpress LLC Supplier No. HY13643810MG
Toltrazuril sulfoxide is an analytical standard corresponding to the short-lived sulfoxide metabolite of toltrazuril. It is intended for research and analytical applications, including metabolic studies, method development, and quantitative analysis of toltrazuril metabolites.
  • Analytical standard suitable for metabolic and pharmacokinetic studies.
  • High purity (99.6%) for reliable analytical results.
  • Provided in small pack sizes for trace-level assays and method validation.
  • Compatible with LC-MS and other chromatographic detection methods.
  • Stable solid form for routine storage and handling.