Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene-2,6-diisocyanate, 97%
Click to view available options
Quantity:
1 g
5 g
Description
Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
Specifications
Specifications
| Melting Point | 13°C |
| Density | 1.226 |
| Boiling Point | 129°C to 133°C (18 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Pungent |
| Refractive Index | 1.5708 |
| Quantity | 5 g |
| UN Number | UN2078 |
| Beilstein | 2211546 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H315-H317-H319-H330-H334-H335-H350 |
| Hazard Statement | GHS H Statement H330-H334-H351-H315-H319-H317-H335 Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P271-P272-P280-P281-P284-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUENE DIISOCYANATE |
| EINECSNumber | 202-039-0 |
| RTECSNumber | CZ6310000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction