Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Toluene, Reagent Grade, 4 Liters
SDP

Catalog No. 50999352
Encompass_Preferred

Product Specifications:

  • Water: 0.03% max
  • Fluorescence: 0.5 pp billion max
  • Evaporation Residue: 0.0004% max
  • UV Cutoff (10 mm path): 285 l (mm)
  • Refractive Index: 1.497 (20°C)
WARNING  This product can expose you to Toluene, a chemical which is known to the State of California to cause female developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Catalog No. 50-999-352 Supplier Research Products International Corp Supplier No. 400015
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Specifications:

  • Water: 0.03% max
  • Fluorescence: 0.5 pp billion max
  • Evaporation Residue: 0.0004% max
  • UV Cutoff (10 mm path): 285 l (mm)
  • Refractive Index: 1.497 (20°C)
WARNING  This product can expose you to Toluene, a chemical which is known to the State of California to cause female developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.