Learn More
Supelco Inc Toluene, 108-88-3, MFCD00008512, 4X4L/CS
Catalog No. NC9034607
Encompass
TOLUENE, >=99.5%, A.C.S. REAGENT, Linear Formula: C6H5CH3, Molecular Weight: 92.14, BP: 110-111 °C(lit.), MP: -93 °C (lit.). Toluene, a flammable liquid with a pungent odor, is widely employed as organic solvent. It is widely used as a precursor for synthesizing benzene and as a solvent in the paint industry. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.
Catalog No. NC9034607 Supplier Supelco Inc Supplier No. 1794184X4L
TOLUENE, >=99.5%, A.C.S. REAGENT, Linear Formula: C6H5CH3, Molecular Weight: 92.14, BP: 110-111 °C(lit.), MP: -93 °C (lit.). Toluene, a flammable liquid with a pungent odor, is widely employed as organic solvent. It is widely used as a precursor for synthesizing benzene and as a solvent in the paint industry. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.