Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187472 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0617 5 mL
11-101-9471 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0617 Supplier MilliporeSigma Supelco Supplier No. 896805ML
Only null left
Add to Cart
Add to Cart

Toluene is an aromatic hydrocarbon, which can be obtained from crude oil via cracking or distillation process.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-88-3
Ignition Point 997°F
Vapor Pressure 26 mmHg (25°C)
Vapor Density 3.2 (vs. air)
Density 0.865 g/mL (at 25°C)
Flash Point 4.0°C
Physical Form Neat
Linear Formula C6H5 CH3
MDL Number MFCD00008512
Melting Point -93°C
Boiling Point 110°C to 111°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 92.14
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.