Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene ACS, MilliporeSigma™

Catalog No. MTX07355 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Poly-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MTX07355 4 L Glass Bottle
MTX0735P4 4 L Poly-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MTX07355 Supplier MilliporeSigma Supplier No. TX07355
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.87g/cm3 at 20°C
Quantity 4 L
Flash Point 4.4°C (39.9°F), Closed cup
Ignition Point 480°C (896°F)
Odor Aromatic
Packaging Glass Bottle
Linear Formula C6H5CH3
UN Number UN 1294
Boiling Point 110.6°C
Solubility Information Insoluble in water
Formula Weight 92.14
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Toluene
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 203-625-9
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.