Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene ACS reagent 99.5200L
Catalog No. 502903686
Encompass
Toluene a flammable liquid with a pungent odor is widely employed as organic solvent. It is widely used as a precursor for synthesizing benzene and as a solvent in the paint industry. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2 by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.
Catalog No. 50-290-3686 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 179418200L
Toluene a flammable liquid with a pungent odor is widely employed as organic solvent. It is widely used as a precursor for synthesizing benzene and as a solvent in the paint industry. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2 by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.