Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOT020020E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOT020020E 20 L
RSOT02004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RSOT020020E Supplier Ricca Chemical Company Supplier No. RSOT020020E4
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

CAS Max % 100
Quantity 20 L
Packaging Metal Can
Residue after Evaporation max. 0.001%
Water max. 0.03%
Boiling Point 111°C
Percent Purity min. 99.5%
Grade ACS
Chemical Name or Material Toluene
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.