Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene anhydrous 99.8
Catalog No. 502903306
Encompass
Toluene a flammable liquid with a pungent odor is widely employed as an organic solvent. It is also used as a precursor for synthesizing benzene.
Catalog No. 50-290-3306 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 24451118L
Toluene a flammable liquid with a pungent odor is widely employed as an organic solvent. It is also used as a precursor for synthesizing benzene.