Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023054 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-3054 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-3054 Supplier MilliporeSigma Supelco Supplier No. RTC00009620ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -93°C (literature)
Density 0.865 g/mL (at 25°C (literature))
Quantity 20 mL
Flash Point 4°C (Not Applicable)
Refractive Index n/D 1.496 (literature)
Molecular Formula C7H8
Vapor Pressure 26 mmHg (25°C)
Linear Formula C6H5CH3
MDL Number MFCD00008512
Synonym Toluene
Boiling Point 110°C to 111°C (literature)
Formula Weight 92.14
Grade Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.