Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, CHROMASOLV™, for analysis of dioxins, furans and PCB, Solstice

Catalog No. 6001704 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-04 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-017-04 Supplier Solstice Supplier No. 344132.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for analysis of dioxins, furans and PCB,≥99.7%

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Color Colorless
Density 0.865g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Vapor Pressure 22 mmHg (20°C), 26 mmHg (25°C)
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Formula Weight 92.14g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV
Chemical Name or Material Toluene
Show More Show Less
Hazard Statement P210-P261-P281-P301 + P310-P331
RTECSNumber XS5250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.