Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, CHROMASOLV™, for HPLC, 99.9%, Solstice

Catalog No. 6001706 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-017-06 1 L Glass Bottle
60-017-08 2.5 L Glass Bottle
60-017-12 6 x 1 L Glass Bottle
60-017-10 4 x 2.5 L Glass Bottle
60-017-11 4 x 4 L Glass Bottle
60-017-05 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-017-06 Supplier Solstice Supplier No. 348661L
Add to Cart
Add to Cart

CHROMASOLV™, for HPLC, 99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -93°C (literature)
Absorbance max. 1.00 at 286 nm, max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 300 nm, max. 0.50 at 288 nm
CAS Max % 100.0000%
Density 0.865 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 4°C
Packaging Glass Bottle
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Boiling Point 110.6°C
Formula Weight 92.14
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Toluene, CHROMASOLV™, for HPLC, 99.9%
Show More Show Less
ShelfLife 1800 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene
EINECSNumber 203-625-9
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.