Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene-d8 99.6 atom D10G
Catalog No. 502897928
Encompass
Toluene-d8 is a deuterated NMR solvent useful in NMR-based research and analyses. Spin-lattice relaxation time (T 1) viscosity and density have been measured over -35 to 200C range. The temperature dependent chemical shifts in the proton NMR spectra of -unsaturated ketones in toluene-d8 has been studied.
Catalog No. 50-289-7928 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 43438810G
Toluene-d8 is a deuterated NMR solvent useful in NMR-based research and analyses. Spin-lattice relaxation time (T 1) viscosity and density have been measured over -35 to 200C range. The temperature dependent chemical shifts in the proton NMR spectra of -unsaturated ketones in toluene-d8 has been studied.