Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene-d8, 99.6%(Isotopic), Thermo Scientific Chemicals

Catalog No. AA3647709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3647709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3647709 Supplier Thermo Scientific Chemicals Supplier No. 03647709
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2037-26-5 | C7H8 | 100.19 g/mol

Toluene is mainly used as a precursor to benzene. Paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, and leather tanners. And also used as an octane booster in gasoline fuels used in internal combustion engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2037-26-5
Molecular Formula C7H8
Molecular Weight (g/mol) 100.19
MDL Number MFCD00044638
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
Synonym toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
PubChem CID 74861
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
SMILES CC1=CC=CC=C1

Specifications

Physical Form Liquid
Quantity 10 g
Density 0.94 g/mL
Flash Point 10°C (50°F)
Odor Aromatic
Melting Point -93°C
Boiling Point 111°C
UN Number UN1294
Solubility Information Soluble in water.0.5g/L (20°C)
Formula Weight 100.19
Percent Purity 99.6%
Chemical Name or Material Toluene-d8, Isotopic
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE
EINECSNumber 218-009-5
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.