Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene-d8 99 atom D10G
Catalog No. 502898841
Encompass
Toluene-d8 is a deuterated derivative of toluene. Pyrolytic decomposition of dilute mixtures (concentration range 5-20ppm) of toluene-d8 in argon behind shock waves has been reported to afford various concentrations of hydrogen and deuterium atoms. This pyrolytic reaction was investigated by resonance absorption spectroscopy. It participates as solvent in the time-course 1H-NMR (Proton Nuclear Magnetic Resonance) spectral evaluation of (E)-dimethyl 2-(ferrocenylethynyl)-3-(4-nitrophenylethynyl)fumarate on irradiation with light at 546 and 578nm.
Catalog No. 50-289-8841 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 15199810G
Toluene-d8 is a deuterated derivative of toluene. Pyrolytic decomposition of dilute mixtures (concentration range 5-20ppm) of toluene-d8 in argon behind shock waves has been reported to afford various concentrations of hydrogen and deuterium atoms. This pyrolytic reaction was investigated by resonance absorption spectroscopy. It participates as solvent in the time-course 1H-NMR (Proton Nuclear Magnetic Resonance) spectral evaluation of (E)-dimethyl 2-(ferrocenylethynyl)-3-(4-nitrophenylethynyl)fumarate on irradiation with light at 546 and 578nm.