Learn More
Cambridge Isotope Laboratories Toluene-d8 (d, 99.5%), 2037-26-5, MFCD00044638, 10x1 g
Catalog No. NC9976515
Encompass_Preferred
Chemical Formula: C6D5CD3, MW: 100.19, 99.5%.
Catalog No. NC9976515 Supplier Cambridge Isotope Laboratories Supplier No. DLM510X1
Chemical Formula: C6D5CD3, MW: 100.19, 99.5%.