Learn More
Cambridge Isotope Laboratories Toluene-d8 (d, 99.5%), 2037-26-5, MFCD00044638, 25 g
Catalog No. NC9267030
Encompass_Preferred
Chemical Formula: C6D5CD3, MW: 100.19, 99.5%.
Catalog No. NC9267030 Supplier Cambridge Isotope Laboratories Supplier No. DLM525
Chemical Formula: C6D5CD3, MW: 100.19, 99.5%.