Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene-d8, for NMR, 99.5+ atom % D

Catalog No. AC321320010
Change view
Click to view available options
Quantity:
1 mL
10 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC321320010 1 mL Glass Bottle
AC321320100 10 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC321320010 Supplier Thermo Scientific Chemicals Supplier No. 321320010
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2037-26-5 | C7H8 | 100.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2037-26-5
Molecular Formula C7H8
Molecular Weight (g/mol) 100.19
MDL Number MFCD00044638
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
Synonym toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
PubChem CID 74861
IUPAC Name 1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
SMILES CC1=CC=CC=C1

Specifications

Physical Form Liquid
Color Colorless
Flash Point 4°C
Refractive Index 1.494
Packaging Glass Bottle
Quantity 1 mL
Deuterium (D) 99.50% min.
Density 0.9400g/mL
Melting Point -84.0°C
Boiling Point 110.0°C
Linear Formula C6D5CD3
Beilstein 05, IV, 797
Formula Weight 100.21
Grade NMR
Chemical Name or Material Toluene-d8, 99.5+ atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-009-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.