Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene-d8, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D

Catalog No. AC464120075
Change view
Click to view available options
Quantity:
7.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464120075 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464120075 Supplier Thermo Scientific Chemicals Supplier No. 464120075
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2037-26-5 | C7H8 | 100.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2037-26-5
Molecular Formula C7H8
Molecular Weight (g/mol) 100.19
InChI Key YXFVVABEGXRONW-JGUCLWPXSA-N
IUPAC Name 1-(²H₃)methyl(²H₅)benzene
SMILES [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])C([2H])([2H])[2H]

Specifications

Quantity 7.5 mL
Flash Point 4°C
Density 0.9400 g/mL
Specific Gravity 0.94
Refractive Index 1.4940
Melting Point -84.0°C
Boiling Point 110.0°C
Linear Formula C6D5CD3
Solubility Information Solubility aq. soln.: almost insoluble (0.5 g/L (at 20°C).)
Formula Weight 100.21
Grade NMR
Chemical Name or Material Toluene-d8
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Highly flammable liquid and vapor.

May be fatal if swallowed and enters airways.

May cause drowsiness or dizziness.

May cause damage to organs through prolonged or repeated exposure if inhaled.

Suspected of
Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

Wash face, hands and any exposed skin thoroughly after handling.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Ground and bond container and receiving equipment
EINECSNumber 218-009-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.