Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Dried, SeccoSolv™, MilliporeSigma™

Catalog No. M1083261001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
150 mL
500 mL
1 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1083261001 1 L Glass Bottle
M1083260161 150 mL Glass Bottle
M1083260500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1083261001 Supplier MilliporeSigma Supplier No. 1.08326.1001

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.87 g/3 (20 °C)
Quantity 1 L
Flash Point 4°C
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 29 hPa (20 °C)
Solubility Information 0.52g/L
Boiling Point 110.6°C (1013 hPa)
Percent Purity >99.995%
Chemical Name or Material Toluene
Show More Show Less
EINECSNumber 203-625-9
RTECSNumber XS5250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.