Learn More
Specifications
Specifications
| Melting Point | -95°C |
| Color | Colorless |
| Density | 0.866g/cm³ |
| Quantity | 4 L |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 29mbar at 20°C |
| Linear Formula | C6H5CH3 |
| Viscosity | 0.6 mPa/s at 20°C |
| Show More |
Safety and Handling
| Hazard Category | Signal Word: Danger |
| Hazard Statement | DANGER! |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.