Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Solstice

Catalog No. 60090594 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1250 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-594 1250 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-594 Supplier Solstice Supplier No. CS347ZA1250
Add to Cart
Add to Cart
Request a Quote

ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
PubChem CID 1140
ChEBI CHEBI:17578

Specifications

Quantity 1250 L
Flash Point 4 °C
Packaging IBC
Physical Form Liquid
Grade ACS
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.