Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Solstice

Catalog No. 60090593 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-593 56 L
60-090-592 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-593 Supplier Solstice Supplier No. CS34756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC, for spectrophotometry, >99.8%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
PubChem CID 1140
ChEBI CHEBI:17578

Specifications

Quantity 56 L
Packaging Returnable Steel Drum
Physical Form Liquid
Refractive Index 1.4963 - 1.4971
Molecular Formula C7H8
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.