Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene HPLC Plus, for HPLC, GC, and residue analysis, >=99.9% | 108-88-3 | MFCD00008512 | 6X1L
Catalog No. NC1496753
Encompass
Toluene HPLC Plus, for HPLC, GC, and residue analysis, >=99.9% | Purity: >=99.9% | Mol Wt: 92.14 | 108-88-3 | MFCD00008512 | 6X1L
Catalog No. NC1496753 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6505796X1L
Toluene HPLC Plus, for HPLC, GC, and residue analysis, >=99.9% | Purity: >=99.9% | Mol Wt: 92.14 | 108-88-3 | MFCD00008512 | 6X1L