Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Laboratory Reagent, ≥99.3%, Solstice

Catalog No. 60045011 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-011 10 L Stainless Steel Can
60-045-012 1 L Glass Bottle
60-045-013 2.5 L Glass Bottle
60-045-016 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-045-011 Supplier Solstice Supplier No. 17996510L
Add to Cart
Add to Cart

Laboratory Reagent,≥99.3%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.865g/mL at 25°C
Quantity 10 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Packaging Stainless Steel Can
Physical Form Liquid
Refractive Index n/D 1.496(lit.)
Vapor Pressure 22mmHg (20°C), 26mmHg (25°C)
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Boiling Point 110.6°C
Formula Weight 92.14g/mol
Percent Purity ≥99.3%
Grade Laboratory Reagent
Chemical Name or Material Toluene
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H361d-H373
DOTInformation class 3 - PG 2 - Toluene
EINECSNumber 203-625-9
RTECSNumber XS5250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.