Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Low water, BakerDRY™, J.T. Baker™

Catalog No. 02004475 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-475 1 L Narrow Mouth Amber Glass Bottle, w/ Septum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-475 Supplier Avantor Supplier No. 936412
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Specifications

CAS 108-88-3
Quantity 1 L
Flash Point 4.00°C
Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Specific Gravity 1L = 0.86kg
Molecular Formula C7H8
MDL Number MFCD00008512
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
PubChem CID 1140
ChEBI CHEBI:17578
Formula Weight 92.14
Grade BakerDRY
Chemical Name or Material Toluene, Low water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.