Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, PRF, Meets requirements of ASTM D2699 and D2700, Solstice

Catalog No. 6001728 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Steel Can
Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-017-28 204 L Steel Drum
60-017-27 19 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-017-28 Supplier Solstice Supplier No. RS347204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

CAS Max % 100.0000%
Density 0.867 g/cm3
Quantity 204 L
Flash Point 4°C
Packaging Steel Drum
UN Number UN1294
Boiling Point 110.6°C
Formula Weight 92.14
Percent Purity >99.5%
Grade Primary Reference Fuel
Chemical Name or Material Toluene, PRF, B&J Brand, Meets requirements of ASTM D2699 and D2700
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.