Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.7% (GC), Solstice

Catalog No. 60046105 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-105 6 x 1 L Glass Bottle
60-046-100 1 L Glass Bottle
60-046-103 4 x 2.5 L Glass Bottle
60-046-101 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-105 Supplier Solstice Supplier No. 322496X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.7% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -93°C
Appearance Colorless liquid
Density 0.865g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Packaging Glass Bottle
Refractive Index n20/D 1.496 to 1.498
Residue after Evaporation 0 to 0.001%
Titratable Acid Free acid: ≤0.001% (as HCl)
Titratable Base Free alkali: ≤0.001% (as NAOH)
Vapor Pressure 22mmHg (20°C), 26mmHg (25°C)
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Boiling Point 110°C to 111°C
Formula Weight 92.14g/mol
Percent Purity ≥99.7% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Toluene, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H361d-H373
DOTInformation Class 3 - PG 2 - Toluene
EINECSNumber 203-625-9
RTECSNumber XS5250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.