Learn More
Aqua Solutions Toluene Reagent, ACS Grade, colorless liquid, 20 L, CAS 108-88-3, minimum assay 99.9%, density 0.8623 g/ml, boiling point 110.60 °C, flash point 4 °C, hazard class 3 (flammable liquid), packaging metal container
SDP
Catalog No. NC2746743
Encompass_Preferred
Toluene Reagent, ACS Grade, colorless liquid, 20 L, CAS 108-88-3, minimum assay 99.9%, density 0.8623 g/ml, boiling point 110.60 °C, flash point 4 °C, hazard class 3 (flammable liquid), packaging metal container
Catalog No. NC2746743 Supplier Aqua Solutions Supplier No. T760120LHP
Toluene Reagent, ACS Grade, colorless liquid, 20 L, CAS 108-88-3, minimum assay 99.9%, density 0.8623 g/ml, boiling point 110.60 °C, flash point 4 °C, hazard class 3 (flammable liquid), packaging metal container