Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene (Scintanalyzed™), Fisher Chemical

Catalog No. T3134 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 4L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
T313-4 Glass Bottle; 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T313-4
Request Bulk or Custom Formats

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 111°C
Melting Point -95°C
Physical Form Liquid
CAS 108-88-3
Background 50 CPM max.
Density 0.866g/cm³
Color Colorless
Quantity 4 L
Flash Point 4°C
Assay Percent Range ≥99.5 %
Molecular Formula C7H8
Linear Formula C6H5CH3
MDL Number MFCD00008512
UN Number 1294
Packaging Glass Bottle
Synonym Methylbenzene, Toluol, Phenyl Methane
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
PubChem CID 1140
ChEBI CHEBI:17578
Formula Weight 92.14
Percent Purity ≥99.5%
Grade Also meets ACS specifications
Water 0.03% max.
Chemical Name or Material Toluene
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.