Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene suitable for HPLC
Catalog No. 502899170
Encompass
Toluene a benzene analog is widely employed as an organic solvent. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2 by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.
Catalog No. 50-289-9170 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348662L
Toluene a benzene analog is widely employed as an organic solvent. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2 by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.