Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene suitable for HPLC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 6485664X4L
Encompass
Toluene a benzene analog is widely employed as an organic solvent. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2 by the denitrifying bacterium Thauera arornatica has been reported. It forms syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.
Catalog No. 50-290-7144