Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Boiling Point | 111°C |
| Grade | Technical |
| Chemical Name or Material | Toluene |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
