Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Technical, Spectrum™ Chemical
SDP

Catalog No. 18614488 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-488 55 gal. Drum
18-614-487 500 mL Amber Glass Bottle
18-614-486 4 L Amber Glass Bottle
18-614-485 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-614-488 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T109255GLBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 108-88-3

Specifications

CAS Max % 1
Quantity 55 gal.
Packaging Drum
Boiling Point 111°C
Grade Technical
Chemical Name or Material Toluene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.