Learn More
Electron Microscopy Sciences Toluidine Blue O, Certified C.I. DcU-10 50 GR
SDP
Catalog No. 5029496
Encompass_Preferred
(Basic blue; Methylene Blue T50 or T Extra)
C₁₅H₁₆N₃SCl F.W. 305.82
Solubility: 3.25% Water; 1.75% Alcohol; 3.5% Cellosolve; 5.5% Glycol; 0.0% Xylene
A general stain for epoxy thick sections.
Catalog No. 50-294-96 Supplier Electron Microscopy Sciences Supplier No. 22050
(Basic blue; Methylene Blue T50 or T Extra)
C₁₅H₁₆N₃SCl F.W. 305.82
Solubility: 3.25% Water; 1.75% Alcohol; 3.5% Cellosolve; 5.5% Glycol; 0.0% Xylene
A general stain for epoxy thick sections.